Learn More
Novacyt - Microgen Bioproducts Path-Chek Salmonella Swab Test
Simple and instant swab test for the detection of Salmonella by direct testing of surfaces.
Brand: Novacyt - Microgen Bioproducts PC020
Additional Details : Weight : 1.40000kg
Description
- Path-Chek Hygiene Test
- Rapid test system designed for the detection of a range of bacteria from work surfaces and the processing environment.
- Simple and instant swab test for the detection of Salmonella by direct testing of surfaces.
Specifications
Salmonella | |
Swab Test | |
100 Tests |