missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX9 Rabbit anti-Human, Mouse, Rat, Clone: 3C2U5, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | PAX9 |
---|---|
Clone | 3C2U5 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18361857
|
Bio-Techne
NBP3-15888-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323534
|
Bio-Techne
NBP3-15888-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAX9 Monoclonal antibody specifically detects PAX9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PAX9 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
paired box 9, paired box gene 9, paired box protein Pax-9, paired domain gene 9, STHAG3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PAX9 (P55771). MEPAFGEVNQLGGVFVNGRPLPNAIRLRIVELAQLGIRPCDISRQLRVSHGCVSKILARYNETGSILPGAIgGSKPRVTTPTVVKHIRTYKQRDPGIFAW | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3C2U5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5083 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title