missing translation for 'onlineSavingsMsg'
Learn More

PCDH11Y Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen PCDH11Y
Concentration 0.18 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899544
View Documents
Proteintech
20069-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16889544
View Documents
Proteintech
20069-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody can recognize PCDH11X and PCDH11Y.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
Specifications

Specifications

PCDH11Y
Western Blot
Unconjugated
Rabbit
Human
Q9BZA7, Q9BZA8
27328, 83259
Peptide
Primary
-20°C
PCDH11Y
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
PCDH PC, PCDH Y, PCDH11, PCDH11X, PCDH11Y, PCDH22, PCDHY, Protocadherin 11, protocadherin 11 Y linked, Protocadherin 22, Protocadherin PC, Protocadherin prostate cancer
PCDH11X, PCDH11Y
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.