Learn More
Brady™ PCR Tube Label for BBP33/i3300/i5300 Printers
The B-492 PCR tube label contains 8 smaller, perforated labels in a single label strip that can be applied on 8 PCR tubes in a rack. It can easily be torn into separate labels thanks to the perforations.
Brand: Brady™ 307018
This item is not returnable.
View return policy
Description
For forations.
Specifications
Permanent Acrylic | |
Matte | |
Polyester (B-492) | |
Labels | |
9.50 mm (W) x 6.35 mm (H) | |
9.50 mm |
White | |
Roll | |
Thermal Transfer | |
Non-sterile | |
6.35 mm | |
9.50 mm |