Learn More
BRAND™ PCR Tube Strips
Extra-thin and even wall thicknesses allow for optimal heat transfer and short cycle times. Smooth shaft surfaces minimize enzyme and nucleic acid binding to the surface ensuring the reaction can run in full.
Brand:  BRAND™ 781283
 View more versions of this product
Product Code. 10789064
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Polypropylene 8 and 12 tube strips
  • A variety of cap styles is available
  • Compatible with leading thermal cyclers with heated lids
  • Provide tight seal in all PCR* cyclers
  • Protect samples from cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Green
125 Pack
Not Included
0.2 mL
Polypropylene
PCR Tube Strip