missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific Pearl™ Adjustable Volume Pipette

The Pearl™ is a single-channel pipette offering adjustable volume and precision liquid handling.

Brand:  Heathrow Scientific HS120106

2147.00 SEK valid until 2025-12-31
Use promo code "24314" to get your promotional price.


 View more versions of this product

Product Code. 16380696

  • 2260.00 SEK / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl™ is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.

  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digital volume display
  • Labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified
  • Autoclavable
Specifications

Specifications

CE
Pearl™ Adjustable Volume Pipette
White
Reusable
HS120106
Pearl
Adjustable Volume Pipette
3-Position, Digit
1 Box
7 cm
2.54 cm
26.04 cm
White
Autoclavable
Low-Force
ABS
1
Non-sterile
1000 to 10,000 μL
Hook
2.75 in.
1.00 in.
10.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heathrow Scientific Pearl™ Adjustable Volume Pipette > Volume Range: 1000 to 10000 μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.