missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pectinase from Aspergillus niger

Brand:  MP Biomedicals™ 0219897910

 View more versions of this product

Product Code. 11416931

  • 290.44 SEK / 10ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.

  • Lyophilized
  • Activity: >1000 u/gm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

>1000 U/g
Used in plant protoplast preparation to digest cell wall prior to organelle isolation
Aspergillus niger
Pectinase
Beige
4°C
10 kU
Lyophilized Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers