Learn More
MP Biomedicals™ Pectinase from Aspergillus niger
Brand: MP Biomedicals™ 0219897980
Additional Details : CAS Number : 9032-75-1 Weight : 0.01000kg
Description
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
- Lyophilized
- Activity: >1000 u/gm
Specifications
>1000 U/g | |
Used in plant protoplast preparation to digest cell wall prior to organelle isolation | |
Aspergillus niger | |
Pectinase |
Beige | |
4°C | |
100 kU | |
Lyophilized Powder |