missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pectinase from Aspergillus niger

Brand:  MP Biomedicals™ 0219897901

Additional Details : CAS Number : 9032-75-1 Weight : 0.01000kg

 View more versions of this product

Product Code. 11496921

  • 244.86 SEK / 1ku
Estimated Shipment: 23-08-2024
to see stock.

Description

Description

Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.

  • Lyophilized
  • Activity: >1000 u/gm

Specifications

Specifications

>1000 U/g
Used in plant protoplast preparation to digest cell wall prior to organelle isolation
Aspergillus niger
Pectinase
Beige
4°C
1 kU
Lyophilized Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers