missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pemetrexed disodium salt 2.5 hydrate, 98%, Thermo Scientific Chemicals
1961.71 SEK - 8776.53 SEK
Chemical Identifiers
CAS | 357166-30-4 |
---|---|
Molecular Formula | C20H19N5Na2O6 . 5/2 H2O |
Molecular Weight (g/mol) | 516.43 |
MDL Number | MFCD13194950 |
InChI Key | NYDXNILOWQXUOF-CETBVFFHNA-L |
Synonym | pemetrexed disodium hemipenta hydrate |
PubChem CID | 131673885 |
IUPAC Name | (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid;molecular hydrogen;sodium;pentahydrate |
SMILES | [Na+].[Na+].NC1=NC2=C(C(CCC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)=CN2)C(=O)N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15365058
|
Thermo Scientific Acros
456180010 |
1 g |
1961.71 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15375058
|
Thermo Scientific Acros
456180050 |
5 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
357166-30-4 | |
516.43 | |
NYDXNILOWQXUOF-CETBVFFHNA-L | |
131673885 | |
[Na+].[Na+].NC1=NC2=C(C(CCC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)=CN2)C(=O)N1 |
C20H19N5Na2O6 . 5/2 H2O | |
MFCD13194950 | |
pemetrexed disodium hemipenta hydrate | |
(2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid;molecular hydrogen;sodium;pentahydrate |
Specifications
357166-30-4 | |
97.5% min. (HPLC) | |
C20H19N5Na2O6 . 5/2 H2O | |
pemetrexed disodium hemipenta hydrate | |
[Na+].[Na+].NC1=NC2=C(C(CCC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)=CN2)C(=O)N1 | |
516.43 | |
516.43 | |
Pemetrexed disodium salt 2.5 hydrate |
Authentic | |
Glass Bottle | |
MFCD13194950 | |
NYDXNILOWQXUOF-CETBVFFHNA-L | |
(2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid;molecular hydrogen;sodium;pentahydrate | |
131673885 | |
98% |
Safety and Handling
GHS H Statement Suspected of causing genetic defects. May damage fertility. May damage the unborn child.
GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RUO – Research Use Only