missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tetrastearate, MP Biomedicals
Click to view available options
Quantity:
1 kg
Specifications
Specifications
CAS | 115-83-3 |
Molecular Formula | C77H148O8 |
MDL Number | MFCD00041922 |
Quantity | 1 kg |
Synonym | pentaerythrityl tetrastearate, pentaerythritol tetrastearate, unii-w9q3dzs0eg, stearic acid, neopentanetetrayl ester, w9q3dzs0eg, 3-octadecanoyloxy-2,2-bis octadecanoyloxymethyl propyl octadecanoate, octadecanoic acid, 2,2-bis 1-oxooctadecyl oxy methyl-1,3-propanediyl ester, pe tetrastearate, pentaerythrittetrastearat |
InChI Key | OCKWAZCWKSMKNC-UHFFFAOYSA-N |
SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)(COC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC |
IUPAC Name | 3-(octadecanoyloxy)-2,2-bis[(octadecanoyloxy)methyl]propyl octadecanoate |
Molecular Weight (g/mol) | 1202.02 |
PubChem CID | 61037 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'