Learn More
Pentanochlor solution, Honeywell Fluka™
100 μg/mL in acetonitrile, analytical standard, reference material
Brand:  Honeywell Fluka R1362-2ML
Product Code. 15989691
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2307-68-8, 75-05-8
2 mL
6°C
Liquid
Glass Bottle
82°C
Herbicides
Analytical Standard
Documents
One moment while we fetch your results.
Product Title
Pentanochlor solution, Honeywell Fluka™ > 2mL