missing translation for 'onlineSavingsMsg'
Learn More

PER3 Rabbit anti-Human, Mouse, Rat, Clone: 9I3I6, Novus Biologicals™

Rabbit Monoclonal Antibody

1905.00 SEK - 4805.00 SEK

Specifications

Antigen PER3
Clone 9I3I6
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18322745
View Documents
Bio-Techne
NBP3-16094-20UL
20 μg
1905.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18317282
View Documents
Bio-Techne
NBP3-16094-100UL
100 μg
4805.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PER3 Monoclonal antibody specifically detects PER3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

PER3
Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila)
Recombinant fusion protein containing a sequence corresponding to amino acids 10-100 of human PER3 (P56645). GRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQIL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
9I3I6
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cardiovascular Biology, Endocrinology
PBS, 0.05% BSA, 50% glycerol, pH7.3
8863
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.