missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin Monoclonal Antibody (5B10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512469
This item is not returnable.
View return policy
Description
MA5-12469 targets Perforin in IHC (P) applications and shows reactivity with Human samples. The MA5-12469 immunogen is recombinant protein corresponding to C-terminal region of human perforin.
Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
Specifications
Perforin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P14222 | |
PRF1 | |
Recombinant protein corresponding to C-terminal region of human perforin | |
RUO | |
5551 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
5B10 | |
Unconjugated | |
PRF1 | |
Cyta; Cytolysin; FLH2; HPLH2; lymphocyte pore forming protein; lymphocyte pore-forming protein; OMAK; OTTHUMP00000019759; P1; perforin 1; perforin 1 (pore forming protein); Perforin1; perforin-1; Pfn; PFN1; Pfp; PGFL; PIGF; PIGF-2; PLGF; pore forming protein; pore-forming protein; PRF1; Prf-1; PRF1 (pore forming protein 1); RATCYTA; RP11-710A11.3 | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Perforin Monoclonal Antibody (5B10), Invitrogen™ > 500 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction