Learn More
Brady™ Permanent Paint
Permanent Paint is ideal for floor marking.
4085.00 SEK
Specifications
Quantity | 6 Pk. |
---|---|
Product Type | Paint |
For Use With (Application) | Floor marking |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11791356
|
Brady™
225005 |
Blue |
4085.00 SEK
1 pack |
In Stock
Log in to see stock. |
|||||
11721366
|
Brady™
225008 |
Black |
4085.00 SEK
1 pack |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
11701366
|
Brady™
225006 |
Green |
4085.00 SEK
1 pack |
In Stock
Log in to see stock. |
|||||
11731366
|
Brady™
225009 |
Gray |
4085.00 SEK
1 pack |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
11711366
|
Brady™
225007 |
Orange |
4085.00 SEK
1 pack |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
11761366
|
Brady™
255355 |
Red |
4085.00 SEK
1 pack |
In Stock
Log in to see stock. |
|||||
11751366
|
Brady™
255354 |
White |
4085.00 SEK
1 pack |
In Stock
Log in to see stock. |
|||||
11741366
|
Brady™
255353 |
Yellow |
4085.00 SEK
1 pack |
In Stock
Log in to see stock. |
|||||
Description
- Floor marking paint.
Specifications
6 Pk. | |
Paint | |
Floor marking |