Learn More
Cytiva Whatman™ PolyCap PES 36
Non-sterile Polycap™ PES (polyethersulfone) capsule filter combines a glass microfiber prefilter and a polyethersulfone membrane prolonging the life of the filter.
Brand:  Cytiva 2642
Additional Details : Weight : 0.51000kg
Product Code. 11369134
  • 3061.00 SEK / Pack of 5
Estimated Shipment: 11-09-2024
to see stock.
Description
Description
  • Allows for large volumes with difficult samples to be filtered easily.
  • Polyethersulfone membrane has low extractables is bio-safe and has excellent flow rates.
  • Ultra-clean containing no surfactant or mold release agents.
  • Housing thermally fused (no glues adhesives or extraneous materials).
  • Integrity-testable by bubble point pressure decay or forward flow methods.
  • Provides highly effective filtration area in a small size. 
Specifications
Specifications
0.2 μm
5Pack
Polycap
5
Non-sterile
36 mm