Learn More
Pyrex™ Borosilicate Glass Pestle
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
180.00 SEK - 230.00 SEK
Specifications
Product Type | Pestle |
---|---|
Material | Borosilicate Glass |
For Use With (Equipment) | Pyrex™ Mortar |
Product Code | Brand | Length (Metric) | Finish | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Finish | Price | Quantity & Availability | |||||
13471869
|
Pyrex
718342R |
110mm | Glazed |
184.00 SEK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
12695327
|
Pyrex
718342 |
110mm | Rough |
180.00 SEK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
12682022
|
Pyrex
718347 |
125mm | Rough |
230.00 SEK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
Description
- Ideal for crushing solids into powders for easier handling
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Available in a rough or glazed finish
- For use with Pyrex™ Mortars (sold separately)
Specifications
Pestle | |
Borosilicate Glass | |
Pyrex™ Mortar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Pestle