Learn More
Motic™ Petri Dish Holder
Firmly holds petri dishes on your microscope stage. Motic™ Petri Dish Holder facilitates reproducible and consistent petri dish viewing.
769.00 SEK
Specifications
Type | Petri Dish Holder |
---|---|
For Use With (Equipment) | AE Series Microscope stages |
Product Code | Brand | Device Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Device Holds | Price | Quantity & Availability | |||||
11834403
|
Motic™
1101000200282 |
Petri Dish (54 mm) |
769.00 SEK
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
15405332
|
Motic™
1101000200272 |
Petri Dish (35 mm) |
769.00 SEK
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
|||||
Description
- For use with AE Series Microscope stages.
- Facilitates reproducible petri dish viewing.
Specifications
Petri Dish Holder | |
AE Series Microscope stages |