missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether 30-40°C, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Amber Glass Bottle
Metal Can
Specifications
Specifications
Melting Point | -130°C |
Boiling Point | 30°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 514mmHg 25 |
IUPAC Name | Petroleum ether |
Formula Weight | 72.15g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction