missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF 184, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent and selective IKKβ inhibitor (IC50 = 37 nM). Displays selectivity over 85 other kinases. Inhibits IL-1β-induced TNF-α production in a steroid-insensitive in vitro model of oxidative stress.
Specifications
CAS | 1187460-81-6 |
Molecular Formula | C32H32ClFN6O4 |
Quantity | 10 mg |
Synonym | 8-[[[5-Chloro-2-[3,4-dimethyl-3,4-bis(hydroxymethyl)-1-pyrrolidinyl]-4-pyridinyl]carbonyl]amino]-1-(4-fluorophenyl)-4,5-dihydro-1H-benz[g]indazole-3-carboxamide |
InChI Key | JUOWWGNRWRLBSV-UHFFFAOYSA-N |
SMILES | CC1(CN(CC1(C)CO)C2=NC=C(C(=C2)C(=O)NC3=CC4=C(CCC5=C4N(N=C5C(=O)N)C6=CC=C(C=C6)F)C=C3)Cl)CO |
IUPAC Name | 8-[[2-[3,4-bis(hydroxymethyl)-3,4-dimethylpyrrolidin-1-yl]-5-chloropyridine-4-carbonyl]amino]-1-(4-fluorophenyl)-4,5-dihydrobenzo[g]indazole-3-carboxamide |
Molecular Weight (g/mol) | 619.094 |
PubChem CID | 76553518 |
Formula Weight | 619.09 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'