Learn More
Cole-Parmer™ PFA Female Threaded Tee Fitting
Threaded fittings offer more permanent installation
1260.00 SEK - 2275.00 SEK
Specifications
Product Type Female Threaded Tee
Description NPT(F) Connection
Autoclavable Not Autoclavable
Material PFA
Sterilization Method Ethylene Oxide
View More Specs
Products 3
Product Code Brand Thread Size Price Quantity & Availability  
Product Code Brand Thread Size Price Quantity & Availability  
11838043
View Documents
Cole-Parmer™
06546-20
1/2 in.
1960.00 SEK
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
11848043
View Documents
Cole-Parmer™
06546-21
3/4 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11828043
View Documents
Cole-Parmer™
06546-19
3/8 in.
1260.00 SEK
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Female Threaded Tee
Not Autoclavable
Ethylene Oxide
Translucent
NPT(F) Connection
PFA
T-shape