missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD Mouse anti-Human, CoraLite 594, Clone: 2A10G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67916-100UL
This item is not returnable.
View return policy
Description
PGD Monoclonal antibody specifically detects PGD in Human samples. It is validated for IFSpecifications
PGD | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Mouse | |
100 μL | |
5226 | |
Store at -20°C | |
Liquid |
Immunofluorescence | |
2A10G4 | |
CoraLite 594 | |
BC018897 | |
Protein G purification | |
Primary | |
Human | |
Antibody | |
IgG1 |