missing translation for 'onlineSavingsMsg'
Learn More
pH 4.01 Calibration Buffer Solution Sachets, Hanna Instruments™ Product Code.: 11687010

pH 4.01 Calibration Buffer Solution Sachets, Hanna Instruments™

Product Code. 11687010
20 mL, 25Pack
Click to view available options
Quantity:
20 mL
Packaging:
25Pack
This item is not returnable. View return policy

Product Code. 11687010

Brand: Hanna Instruments HI70004C

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Premium quality pH 4.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis that is ideal or users that do not calibrate frequently.

  • Premium quality pH 4.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
  • Temperature chart of the actual pH value at various temperatures printed on each sachet
  • pH 4.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware 
  • In a temperature controlled environment monitored with certified thermometers
  • Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability

Specifications

Color Undesignated
pH 4.01
Physical Form Liquid
Quantity 20 mL
Formulation Aqueous Buffer Solution
Odor Odorless
Packaging 25Pack
Traceability to NIST Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH 4.01 Calibration Buffer Solution Sachets, Hanna Instruments™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.