Learn More
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
Brand: MP Biomedicals 0215612850
Description

Specifications
>99% | |
100929-99-5 | |
Store at 0°C. | |
519.471 | |
50mg |
0.854 | |
White | |
Powder | |
Phe-Arg-β-naphthylamide dihydrochloride |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.