missing translation for 'onlineSavingsMsg'
Learn More

Phenol-d1, 95% (Isotopic), Thermo Scientific™

1276.44 SEK - 5028.00 SEK

Chemical Identifiers

CAS 1003-66-3
Molecular Formula C6H5DO
Molecular Weight (g/mol) 95.12
MDL Number MFCD01075468
InChI Key ISWSIDIOOBJBQZ-DYCDLGHISA-N
Synonym phenol-d, phenol-d1, o-2h phenol, phenol-od, 2 h oxy benzene
PubChem CID 12205409
IUPAC Name deuteriooxybenzene
SMILES [2H]OC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437987
View Documents
Thermo Scientific Alfa Aesar
H51100.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15447987
View Documents
Thermo Scientific Alfa Aesar
H51100.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1003-66-3
95.12
ISWSIDIOOBJBQZ-DYCDLGHISA-N
12205409
[2H]OC1=CC=CC=C1
C6H5DO
MFCD01075468
phenol-d, phenol-d1, o-2h phenol, phenol-od, 2 h oxy benzene
deuteriooxybenzene

Specifications

1.082
1003-66-3
182°C
MFCD01075468
phenol-d, phenol-d1, o-2h phenol, phenol-od, 2 h oxy benzene
[2H]OC1=CC=CC=C1
95.12
95.12
Phenol-d1, Isotopic
Air and Light
40°C to 42°C
C6H5DO
UN1671
ISWSIDIOOBJBQZ-DYCDLGHISA-N
deuteriooxybenzene
12205409
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318-H341-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H335-H341-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID

missing translation for 'tsca' : No

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.