missing translation for 'onlineSavingsMsg'
Learn More
Phenol Red Solution R2, Reagecon™ Product Code.: 11404719

Phenol Red Solution R2, Reagecon™
BIOPHARMA

Product Code. 11404719 Shop All Reagecon Products
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 11404719

Brand: Reagecon 1063603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Red
Quantity 500 mL
compliance-icons
Product Identifier
  • PHENOL RED SOLUTION R2
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, sodiumhydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenol Red Solution R2, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.