missing translation for 'onlineSavingsMsg'
Learn More

Phenyl disulfide, 99%, Thermo Scientific Chemicals

536.05 SEK - 1779.22 SEK

Chemische Identifikatoren

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.33
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Packaging Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Packaging Preis Menge & Verfügbarkeit  
10052050
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific Acros
173710500
50 g Plastic bottle
536.05 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
15790819
missing translation for 'viewDocuments'
Thermo Scientific Acros
173712500
250 g Plastic bottle
1779.22 SEK
250g
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

882-33-7
218.33
GUUVPOWQJOLRAS-UHFFFAOYSA-N
13436
C1=CC=C(C=C1)SSC2=CC=CC=C2
C12H10S2
MFCD00003065
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
(phenyldisulfanyl)benzene

Spezifikation

882-33-7
White
Authentic
C12H10S2
MFCD00003065
05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
GUUVPOWQJOLRAS-UHFFFAOYSA-N
(phenyldisulfanyl)benzene
13436
99%
Phenyl disulfide, 99%
58°C to 61°C
191°C to 192°C (15.0 mmHg)
98.5% min. (HPLC)
(C6H5S-)2
06, 323
15, 3355
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
C1=CC=C(C=C1)SSC2=CC=CC=C2
218.33
218.33
Crystalline Solid
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 212-926-4

missing translation for 'rtecsNumber' : SS6825000

missing translation for 'tsca' : TSCA

Produktidentifikator
  • Phenyl disulfide

Signalwort
  • Warning

Gefahrenkategorie
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Gefahrenhinweis
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Sicherheitshinweis
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

RUO – Research Use Only