missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF10 Mouse anti-Human, Clone: PCRP-PHF10-2A10, Novus Biologicals™
Mouse Monoclonal Antibody
2470.00 SEK - 4910.00 SEK
Specifications
Antigen | PHF10 |
---|---|
Clone | PCRP-PHF10-2A10 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18319614
|
Bio-Techne
NBP3-13750-20UG |
20 μg |
2470.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18322123
|
Bio-Techne
NBP3-13750-100UG |
100 μg |
4910.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHF10 Monoclonal antibody specifically detects PHF10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
PHF10 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Q8WUB8 | |
55274 | |
IgG2a | |
Protein A or G purified |
PCRP-PHF10-2A10 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
BAF45a, BAF45APHD zinc finger protein XAP135, BRG1-associated factor 45a, FLJ10975, MGC111009, PHD finger protein 10, XAP135BRG1-associated factor, 45-KD, A | |
Recombinant full-length humanPHF10protein (Uniprot: Q8WUB8) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title