missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Phynomic X-Foam HV Safety Gloves

Product Code. 17220371
Click to view available options
Size:
6
7
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

A seamless break section integrated into the fingers significantly reduces the risk of serious hand injuries when using manually operated screw drivers. Aqua-polymer foam coating on the palm and fingertips enhances grip capabilities.

  • Lightweight construction acts as a second-skin to maintain a high level of feeling
  • Aqua-polymer foam coating is moisture repellent and flexible
  • Breathable materials help prevent excessive perspiration
  • Seamless break section integrated into the fingers helps reduce serious hand injuries
  • Should the glove/finger become trapped in or near a piece of machinery, simply detach and tear along the perforated seam
  • Enhanced grip in dry and slightly damp applications
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Orange, Gray
Material Polyamide, Elastane
Size 8
EN388 Standard 31X1X
Gender Unisex
Glove Cuff Style Knit
Series Phynomic X-Foam HV
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'