missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, 30% Water, Extra Pure, Scharlab

Product Code. 11338654
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent and in chromatography.

  • Synonyms: 2,4,6-Trinitrophenol
  • Molecular Formula: C6H3N3O7
  • Molar Mass: 229.11g/mol
  • CAS: 88-89-1
  • EIN ECS No.: 201-865-9
  • identity (IR-spectrum): Passes test
  • Slightly Soluble in Water (Insoluble and Resinous Matter: max. 0.1%)
  • Chlorides (Cl): max. 0.005%
  • Sulfates (SO4) : max. 0.5%
  • Residue on Ignition: max. 0,1 %

Specifications

CAS 88-89-1
Melting Point 121.4°C
Flash Point 150°C
Ignition Point 300°C
Molecular Formula C6H3N3O7
Quantity 500 g
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,4,6-trinitrophenol
Molecular Weight (g/mol) 229.104
PubChem CID 6954
ChEBI CHEBI:46149
Formula Weight 229.11g/mol
Grade Extra Pure
Concentration or Composition (by Analyte or Components) 98% min assay (acidimetric, on dried sample)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Piric acid, extra pure (with approx. 30% H2O)
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : 2, 4, 6-trinitrophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.