missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picric acid, EssentQ, 70% in water, Scharlab
Click to view available options
Quantity:
250 g
Specifications
Specifications
CAS | 88-89-1 |
Molecular Formula | C6H3N3O7 |
InChI Key | OXNIZHLAWKMVMX-UHFFFAOYSA-N |
SMILES | OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O |
IUPAC Name | 2,4,6-trinitrophenol |
Molecular Weight (g/mol) | 229.10 |
Grade | EssentQ |
Quantity | 250 g |
Safety and Handling
Product Identifier
- Picric acid,extra pure(with approx 30% H2O)
- Danger
- Flammable solid Category 2
- Acute toxicity Category 3
- H228-Flammable solid.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- EUH001-Explosive when dry.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : 2, 4, 6-trinitrophenol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction