Learn More
Pinoresinol Diglucoside, MedChemExpress
MedChemExpress Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.
Brand: Medchem Express HY-N0657
This item is not returnable.
View return policy
Description
Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.Specifications
Pinoresinol Diglucoside | |
Off-White | |
C32H42O16 | |
DMSO : 100 mg/mL (146.48 mM; Need ultrasonic) | |
682.67 | |
98.0% | |
Solid |
63902-38-5 | |
Research | |
50 mg | |
COC(C=C1[C@@H]2[C@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C=C4)([H])[C@]3([H])CO2)=C(C=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO | |
682.67 | |
Research |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.