Learn More
Pinoresinol Diglucoside, MedChemExpress
MedChemExpress Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.
Brand:  Medchem Express HY-N0657
Encompass_Preferred
 View more versions of this product
Product Code. 16444867
  • 5274.87 SEK / 50mg
Explore more special offers
Description
Description
Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.
Specifications
Specifications
Pinoresinol Diglucoside
Off-White
C32H42O16
DMSO : 100 mg/mL (146.48 mM; Need ultrasonic)
682.67
98.0%
Solid
63902-38-5
Research
50 mg
COC(C=C1[C@@H]2[C@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C=C4)([H])[C@]3([H])CO2)=C(C=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO
682.67
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.