Learn More
VITLAB™ Pipette Fillers
Measure the volume of liquids using Vitlab™ Pipette Filters. These filters draw liquid into the pipette.
Brand: VITLAB™ 324594
Additional Details : Weight : 0.02000kg
Description
- Slow rotation of the actuator-wheels draws liquid into the pipette
- Pressing air bleed valve automatically empties the pipette without returning the piston
- Green, Blue and Red available
Specifications
2mL Pipettes, Glass and Plastic Pipettes |