Learn More
Hamilton™ SoftGrip™ Multi-Channel Stand
Providing stability and comfort.
Brand: Hamilton™ 34195
Additional Details : Weight : 1.90400kg
Description
- For use with SoftGrip multi-channel pipettors
- Maximum comfort for left- or right-hand use
- Holds both 8- and 12-channel pipettors
Specifications
8/12-Channel SoftGrip Microlitre Multi-Channel Pipette |