missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Pippette Filter Tip Box with Lid
The Pipette Filter Tip Box is designed to store racked pipette filter tips.
490.00 SEK
Specifications
Includes | Box with Lid |
---|---|
Autoclavable | Not Autoclavable |
Holds | Pipette Tips |
Material | Polypropylene |
Sterility | Non-sterile |
Description
Empty PP boxes with snap-on lid for pipette filter tips.Specifications
Box with Lid | |
Pipette Tips | |
Non-sterile | |
12.52 cm |
Not Autoclavable | |
Polypropylene | |
Pipette Filter Tips |