Learn More
Kern Sohn Plastic Box for Weight Set
Plastic case for individual weight set.
Brand:  Kern Sohn 347-030-400
Encompass_Preferred
 View more versions of this product
Product Code. 17342263
  • 22.52 SEK / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Plastic
  • For button/compact weight design
Specifications
Specifications
Case
Plastic
Class F1 and M3 Weight
1 g to 5 g weights