Learn More
Kartell™ plastilab™ Volumetric Flask with Stopper
These plastilab™ flasks create no meniscus which simplifies the process of reading final volumes. Calibrated gravimetrically at 20°C, the translucency of polypropylene is enhanced when it comes in contact with liquid. Thin walls eliminate distortion, and a thin neck design makes final readings precise. This Volumetric Flask is autoclavable, chemically non-absorbent, safe for use with food, and includes stoppers.
185.00 SEK
Specifications
Color Clear
Material Polypropylene
Autoclavable Autoclavable
View More Specs
Products 1
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11348533
View Documents Promotion Details
Kartell™
0148600
100 mL
185.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-09-2024
to see stock.
Add to basket
Description
Description
These plastilab™ flasks create no meniscus which simplifies the process of reading final volumes. Calibrated gravimetrically at 20°C, the translucency of polypropylene is enhanced when it comes in contact with liquid. Thin walls eliminate distortion, and a thin neck design makes final readings precise. This Volumetric Flask is autoclavable, chemically non-absorbent, safe for use with food, and includes stoppers.
  • Calibrated gravimetrically at 20°C
  • Made from chemically resistant polypropylene
  • Thin walls eliminate distortion
  • Thin neck makes final reading precise
  • Suitable for contact with food products
  • Autoclavable
Specifications
Specifications
Clear