Learn More
Fisherbrand™ Plain Plates with 2mm Spacers
Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System.
Brand:  Fisherbrand™ VS20PGS2
Product Code. 11824542
  • 523.00 SEK / Pack of 2
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System
Specifications
Specifications
Verti-Gel Maxi 2-Gel Vertical Gel Electrophoresis System
Plain Plates with 2 mm Spacers