Learn More
Eppendorf™ Platforms for New Brunswick™ I26/I26R Shaker
Dedicated platform to fit I26/I26R shakers.
16340.10 SEK - 19502.70 SEK
Product Code | Brand | No. of Positions | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Holds | Price | Quantity & Availability | ||||
11652764
|
Eppendorf™
M1324-9908 |
15 | 1 L Erlenmeyer Flasks |
17288.88 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11662764
|
Eppendorf™
M1324-9909 |
12 | 2 L Erlenmeyer Flasks |
16340.10 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
11632764
|
Eppendorf™
M1324-9906 |
40 | 250 mL Erlenmeyer Flasks |
19502.70 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
Description
- Holds Erlenmeyer flasks or Fernbach
- Dedicated platform
Specifications
Innova™ I26/I26R |