Learn More
Eppendorf™ Platforms for New Brunswick™ Innova™ 44/44R Shakers
Dedicated platform to fit 44/44R shakers.
Brand:  Eppendorf™ M1282-9909
 View more versions of this product
Product Code. 11369994
  • 16867.20 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
  • Holds Erlenmeyer flasks or Fernbach
  • Dedicated platform
  • Aluminum
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 L Erlenmeyer Flasks
Aluminum
Innova™ 44/44R
12