Learn More
Eppendorf™ Platforms for New Brunswick™ S41i Incubator Shaker
Interchangeable, dedicated platform for the S41i.
10311.08 SEK - 11279.94 SEK
Specifications
For Use With (Equipment) New Brunswick™ S41i
View More Specs
Products 7
Product Code Brand No. of Positions Holds Price Quantity & Availability  
Product Code Brand No. of Positions Holds Price Quantity & Availability  
13438686
View Documents
Eppendorf™
M1334-9921
32 125 mL Erlenmeyer Flasks
11279.94 SEK
Each
In Stock
to see stock.
Add to basket
13448686
View Documents
Eppendorf™
M1334-9922
24 250 mL Erlenmeyer Flasks
10647.42 SEK
Each
In Stock
to see stock.
Add to basket
13458686
View Documents
Eppendorf™
M1334-9923
15 500 mL Erlenmeyer Flasks
10451.64 SEK
Each
In Stock
to see stock.
Add to basket
13468686
View Documents
Eppendorf™
M1334-9924
11 1 L Erlenmeyer Flasks
10311.08 SEK
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
13478686
View Documents
Eppendorf™
M1334-9925
6 2 L Erlenmeyer Flasks
10311.08 SEK
Each
In Stock
to see stock.
Add to basket
13488686
View Documents
Eppendorf™
M1334-9926
3 2.8 L Erlenmeyer Flasks
10311.08 SEK
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
13498686
View Documents
Eppendorf™
M1334-9927
3 4 L Erlenmeyer Flasks
10311.08 SEK
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Dedicated platform
  • Interchangeable
Specifications
Specifications
New Brunswick™ S41i