Learn More
Platinum Ruthenium foil, 0.05mm (0.002in) thick, Thermo Scientific Chemicals
2740.00 SEK - 21912.00 SEK
Specifications
Chemical Name or Material | Platinum Ruthenium |
---|---|
Color | Silver to Gray |
Appearance | Silver-gary foil |
Molecular Formula | Pt:Ru; 95.2:4.8 |
MDL Number | MFCD00798552 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11345427
|
Thermo Scientific Alfa Aesar
042686.FF |
25 x 25 mm |
2740.00 SEK
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356139
|
Thermo Scientific Alfa Aesar
042686.FI |
50 x 50 mm |
8625.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11335427
|
Thermo Scientific Alfa Aesar
042686.GH |
100 x 100 mm | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Ruthenium | |
Silver-gary foil | |
MFCD00798552 | |
Foil |
Silver to Gray | |
Pt:Ru; 95.2:4.8 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures