Learn More
Memmert™ Plug-In Tube Extension for UF110plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ UF110plusB29718
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
UF110plus Oven | |
Plug-in Tube Extension | |
O.D. 60.3 mm, I.D. 57 mm |
Product Content Correction
Product Title
Memmert™ Plug-In Tube Extension for UF110plus, Straight > For Use With: UF110plus
Spot an opportunity for improvement?Share a Content Correction