Learn More
Kartell™ Pluripet II Variable Volume Micropipette
Ensures a high degree of precision and accuracy combined with a lower cost
2165.00 SEK
Specifications
Autoclavable Autoclavable
Product Line Pluripet II
Type Micropipette
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10315303
View Documents Promotion Details
Kartell™
1319000
0.5, 5.0, 10.0 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10561125
View Documents Promotion Details
Kartell™
1322000
100, 500, 1000 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • The particularly light and balanced body makes the handle comfortable
  • Ideal for dispensing large volumes of rather thick and viscous aqueous fluids
  • Supplied complete with adhesive support for attachment to the laboratory bench
  • Individually calibrated according to the ISO8655 standards, the new international standards for piston- driven volumetric appliances; EC/IVD- compliant
  • Made of high quality plastic and equipped with steel piston for prolonged use over time
  • The desired volume can be set by turning the emptying button; it proves easy and safe thanks to the snap selection
  • The large display ensures simple reading reading of the set volume value
  • The button of each micropipetta has a specific colour so identification of the usable volume interval is simple
  • The tip holder cone can house a wide variety of tips
  • The recalibration device ensures quick and precise recalibrations
  • The robust structure of the body ensures high mechanical and chemical resistances, making them totally autoclavable at +121°C for 20 minutes
  • Specifications
    Specifications
    Autoclavable
    Pluripet II
    Micropipette