missing translation for 'onlineSavingsMsg'
Learn More

PMEL17/SILV Mouse anti-Human, Clone: PMEL/2038, Novus Biologicals™

Mouse Monoclonal Antibody

2450.00 SEK

Specifications

Antigen PMEL17/SILV
Clone PMEL/2038
Concentration 0.2 mg/ml
Dilution Western Blot 2-4 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18337422
View Documents
Bio-Techne
NBP3-07599-20UG
20 μg
2450.00 SEK
20µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

PMEL17/SILV
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Human
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17
A recombinant fragment (around aa 376-502) of human PMEL17/SILV protein (exact sequence is proprietary)
Primary
Store at 4°C.
PMEL/2038
Western Blot 2-4 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
6490
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.