Learn More
PNLIPRP1 Mouse anti-Human, Unconjugated, Clone: 1F11E6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67603-1-IG-20UL
 View more versions of this product
Product Code. 16840326
  • 1656.73 SEK / 20µL
Explore more special offers
Description
Description
The PNLIPRP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PNLIPRP1. This antibody recognizes human antigen. The PNLIPRP1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PNLIPRP1
Monoclonal
500 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1F11E6
Unconjugated
20 μL
5407
IgG2b