missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPLA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25469-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the patatin-like family of phospholipases. The encoded enzyme has both triacylglycerol lipase and transacylase activities and may be involved in adipocyte triglyceride homeostasis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome Y.Specifications
PNPLA4 | |
Polyclonal | |
Unconjugated | |
PNPLA4 | |
DXS1283E, GS2, IPLA2 ETA, PNPLA4, Protein GS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8228 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41247 | |
PNPLA4 | |
PNPLA4 Fusion Protein Ag22091 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |