missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Glutamic Acid, MW 50,000-100,000
9045.00 SEK - 13925.00 SEK
Specifications
Purity | 95% |
---|---|
Content And Storage | 0°C |
Format | Powder |
For Use With (Application) | Used in the construction of Polypeptide based Copolymer membranes, micelles and nanoparticles |
Molecular Weight (g/mol) | 50000 to 100000 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447162
|
MP Biomedicals™
0215191891 |
1000 mg |
13925.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437162
|
MP Biomedicals™
0215191890 |
500 mg |
9045.00 SEK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
95% | |
Powder | |
50000 to 100000 |
0°C | |
Used in the construction of Polypeptide based Copolymer membranes, micelles and nanoparticles | |
Poly-L-Glutamic Acid from Synthetic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title