Learn More
Ansell Polyethylene Shoecovers
Shop All Ansell Products
Click to view available options
Dimensions (L x W):
15 x 36cm
Description
- Cool, light and comfortable to wear
- Manufactured from blue chlorinated polyethylene
- With elastic opening
Specifications
Specifications
Material | Polyethylene |
Color | Blue |
Model | SC-36BCPE |
Packaging Quantity | 1/Pk. |
Dimensions (L x W) | 15 x 36cm |
Product Type | Shoe Covers |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Ansell Polyethylene Shoecovers >
Spot an opportunity for improvement?Share a Content Correction