missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Polyethylene Coveralls

Product Code. 11856910
Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Product Type Single-use Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Sterility Non-sterile
Resistance Tears
Certifications/Compliance (Europe) EC Category PPE, Cat III to Directive 89/686/EEC
For Use With (Application) Chemical Industries, Maintenance, Medical and Pharmaceuticals, Spray Painting, Automotive
Weight (Metric) 55 g/m2
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'